ENAH3155241B Enamine

methyl 5-bromo-2,3-dihydro-1H-indole-3-carboxylate hydrochloride

  • MDL number MFCD31794609
  • Cas Number : 2287283-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744471
Availability details for
Quantity
  • Availability
  • Add to Cart