ENAH31552422 Enamine

9-azatricyclo[9.4.0.0,3,8]pentadeca-1(15),3,5,7,11,13-hexaene

  • MDL number MFCD03723999
  • Cas Number : 449-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746539
Availability details for
Quantity
  • Availability
  • Add to Cart