ENAH31552428 Enamine

2-bromo-1-(5-methylfuran-2-yl)ethan-1-one

  • MDL number MFCD09261158
  • Cas Number : 1634-53-3
  • Molecular Weight : 203.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-65
Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6746591

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6746591
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart