ENAH31552429 Enamine

1-bromocyclopentane-1-carbaldehyde

  • MDL number MFCD24682371
  • Cas Number : 408330-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744494
Availability details for
Quantity
  • Availability
  • Add to Cart