ENAH31552437 Enamine

1-(ethylamino)butan-2-ol

  • MDL number MFCD12801538
  • Cas Number : 68058-17-3
  • Molecular Weight : 117.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745167
Availability details for
Quantity
  • Availability
  • Add to Cart