ENAH3155243A Enamine

(2E)-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylbut-2-enoic acid

  • MDL number MFCD31794999
  • Cas Number : 2164014-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746640
Availability details for
Quantity
  • Availability
  • Add to Cart