ENAH31552443 Enamine

(1S)-1-(4-methoxy-3-nitrophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD24428587
  • Cas Number : 2287247-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746729
Availability details for
Quantity
  • Availability
  • Add to Cart