ENAH3155244C Enamine

6-amino-5H,6H,7H,8H-imidazo[1,2-a]pyridine-2-carboxylic acid dihydrochloride

  • MDL number MFCD31794761
  • Cas Number : 2287314-19-4
  • Molecular Weight : 254.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746762
Availability details for
Quantity
  • Availability
  • Add to Cart