ENAH31552453 Enamine

(1R,5R)-6,6-dimethylspiro[bicyclo[3.1.1]heptane-2,1'-cyclopropane]-3-one

  • MDL number MFCD31795017
  • Cas Number : 1932245-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746792
Availability details for
Quantity
  • Availability
  • Add to Cart