ENAH31552454 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetyl chloride

  • MDL number MFCD31794763
  • Cas Number : 17686-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746816
Availability details for
Quantity
  • Availability
  • Add to Cart