ENAH3155245E Enamine

tert-butyl N-[1-(4-cyanopyridin-2-yl)piperidin-4-yl]carbamate

  • MDL number MFCD09607549
  • Cas Number : 848500-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745299
Availability details for
Quantity
  • Availability
  • Add to Cart