ENAH31552461 Enamine

3,3-dimethoxy-2,2,4,4-tetramethylcyclobutan-1-one

  • MDL number MFCD01928641
  • Cas Number : 133729-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745324
Availability details for
Quantity
  • Availability
  • Add to Cart