ENAH31552463 Enamine

(4-amino-3-methylphenyl)methanesulfonamide

  • MDL number MFCD24627310
  • Cas Number : 915192-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748115
Availability details for
Quantity
  • Availability
  • Add to Cart