ENAH31552466 Enamine

7-tert-butyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD31794614
  • Cas Number : 1695688-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745346
Availability details for
Quantity
  • Availability
  • Add to Cart