ENAH31552469 Enamine

5-[(tert-butoxy)carbonyl]-8-oxa-5-azaspiro[3.5]nonane-7-carboxylic acid

  • MDL number MFCD31794442
  • Cas Number : 2287310-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745354
Availability details for
Quantity
  • Availability
  • Add to Cart