ENAH3155246F Enamine

5-(3-bromophenyl)-1,2,4-triazin-3-amine

  • MDL number MFCD06739755
  • Cas Number : 886497-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745361
Availability details for
Quantity
  • Availability
  • Add to Cart