ENAH31552475 Enamine

2-[(2-aminoethyl)amino]-5-nitrobenzoic acid hydrochloride

  • MDL number MFCD31794805
  • Cas Number : 148819-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743105
Availability details for
Quantity
  • Availability
  • Add to Cart