ENAH31552479 Enamine

4-(aminomethyl)-2-methyl-1lambda6-thiane-1,1-dione hydrochloride

  • MDL number MFCD31794563
  • Cas Number : 2287271-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743276
Availability details for
Quantity
  • Availability
  • Add to Cart