ENAH38A2B97E Enamine

1-(5-fluoropyridin-2-yl)cyclopropane-1-carbonitrile

  • MDL number MFCD23134886
  • Cas Number : 1302580-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130857
Availability details for
Quantity
  • Availability
  • Add to Cart