ENAH38A2BC5A Enamine

1-(1,2-oxazol-3-yl)ethan-1-one

  • MDL number MFCD16619202
  • Cas Number : 88511-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-261519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261519
Availability details for
Quantity
  • Availability
  • Add to Cart