ENAH38A2BD52 Enamine

tert-butyl 4-iodo-2,3-dihydro-1H-pyrrole-1-carboxylate

  • MDL number MFCD28123740
  • Cas Number : 1196146-93-6
  • Molecular Weight : 295.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-658723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-658723
Availability details for
Quantity
  • Availability
  • Add to Cart