ENAH38A2BD54 Enamine

(2-aminoethyl)(ethyl)[(3-fluorophenyl)methyl]amine

  • MDL number MFCD14627423
  • Cas Number : 1247567-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-903270

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-903270
pictograms GHS05,GHS07
hstatements H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart