ENAH38A2BD55 Enamine

6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-amine

  • MDL number MFCD23789735
  • Cas Number : 1180519-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-908433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-908433
Availability details for
Quantity
  • Availability
  • Add to Cart