ENAH38A2BD71 Enamine

3-amino-1H,5H,6H,7H,7aH-pyrrolo[1,2-c]imidazol-1-one

  • MDL number MFCD19690782
  • Cas Number : 948915-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-864944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-864944
Availability details for
Quantity
  • Availability
  • Add to Cart