ENAH38A2BD92 Enamine

2-cyclopentylcyclopentan-1-one

  • MDL number MFCD00051419
  • Cas Number : 4884-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1246886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1246886
Availability details for
Quantity
  • Availability
  • Add to Cart