ENAH38A2BDA6 Enamine

methyl 1H,4H,6H,7H-pyrano[4,3-c]pyrazole-3-carboxylate

  • MDL number MFCD20921989
  • Cas Number : 2133444-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1264210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1264210
Availability details for
Quantity
  • Availability
  • Add to Cart