ENAH38A2BDE2 Enamine

7-methylpyrazolo[1,5-a]pyridine-3-carboxylic acid

  • MDL number MFCD16620169
  • Cas Number : 16205-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1665669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1665669
Availability details for
Quantity
  • Availability
  • Add to Cart