ENAH38A2BDF9 Enamine

ethyl 4-oxohexanoate

  • MDL number MFCD00068986
  • Cas Number : 3249-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102899
Availability details for
Quantity
  • Availability
  • Add to Cart