ENAH38A2BE78 Enamine

7-chloro-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD09701288
  • Cas Number : 658683-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2995455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2995455
Availability details for
Quantity
  • Availability
  • Add to Cart