ENAH38A2C06E Enamine

6-bromo-3H,4H-pyrido[2,3-d]pyrimidin-4-one

  • MDL number MFCD14546963
  • Cas Number : 155690-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113860
Availability details for
Quantity
  • Availability
  • Add to Cart