ENAH38A2CC2D Enamine

1,6-diamino-4-(trifluoromethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD31810947
  • Cas Number : 2135614-60-5
  • Molecular Weight : 193.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739871
Availability details for
Quantity
  • Availability
  • Add to Cart