ENAH38A2CC2E Enamine

tert-butyl 2,3-dimethoxymorpholine-4-carboxylate

  • MDL number MFCD31810948
  • Cas Number : 2305253-67-0
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739880
Availability details for
Quantity
  • Availability
  • Add to Cart