ENAH38A2CC32 Enamine

tert-butyl 1-(methylamino)-2,3,4,5-tetrahydro-1H-3-benzazepine-3-carboxylate

  • MDL number MFCD31810688
  • Cas Number : 2305252-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740659
Availability details for
Quantity
  • Availability
  • Add to Cart