ENAH38A2CC34 Enamine

5-fluoro-1H-indole-3-carbohydrazide

  • MDL number MFCD31810690
  • Cas Number : 1997-83-7
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740885
Availability details for
Quantity
  • Availability
  • Add to Cart