ENAH38A2CC40 Enamine

tert-butyl (3S)-4-(chlorosulfonyl)-3-methylpiperazine-1-carboxylate

  • MDL number MFCD31810711
  • Cas Number : 2305185-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746464

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6746464
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart