ENAH38A2CC41 Enamine

4-chloro-6-methyl-1-phenyl-1,2-dihydropyrimidin-2-one

  • MDL number MFCD31810963
  • Cas Number : 2305252-02-0
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746514
Availability details for
Quantity
  • Availability
  • Add to Cart