ENAH38A2CC44 Enamine

4-(tert-butoxy)butan-1-amine hydrochloride

  • MDL number MFCD31810716
  • Cas Number : 2305252-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748318
Availability details for
Quantity
  • Availability
  • Add to Cart