ENAH38A2CC45 Enamine

5-[(1R)-1-aminoethyl]pyridin-2-ol dihydrochloride

  • MDL number MFCD31810720
  • Cas Number : 2305184-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748334
Availability details for
Quantity
  • Availability
  • Add to Cart