ENAH38A2CC49 Enamine

(1s,3s)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-1-(trifluoromethyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD31810731
  • Cas Number : 2305253-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748538
Availability details for
Quantity
  • Availability
  • Add to Cart