ENAH38A2CC4C Enamine

(1r,3r)-3-chloro-N'-hydroxycyclobutane-1-carboximidamide

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748651
Availability details for
Quantity
  • Availability
  • Add to Cart