ENAH38A2CC4F Enamine

2,3-difluoro-2,3-dimethylbutanenitrile

  • MDL number MFCD31810740
  • Cas Number : 2305252-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748683
Availability details for
Quantity
  • Availability
  • Add to Cart