ENAH38A2CC50 Enamine

3-(chloromethyl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD31810741

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748685
Availability details for
Quantity
  • Availability
  • Add to Cart