ENAH38A2CC52 Enamine

tert-butyl 5,7-dimethyl-1,4-diazepane-1-carboxylate

  • MDL number MFCD31810743
  • Cas Number : 194032-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748778
Availability details for
Quantity
  • Availability
  • Add to Cart