ENAH38A2CC58 Enamine

tert-butyl 4-[imino(methyl)oxo-lambda6-sulfanyl]-1,4-diazepane-1-carboxylate hydrochloride

  • MDL number MFCD31810759
  • Cas Number : 2305255-79-0
  • Molecular Weight : 313.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6748925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748925
Availability details for
Quantity
  • Availability
  • Add to Cart