ENAH38A2CC5D Enamine

tert-butyl 1-(2-aminoethyl)-1,2,3,4-tetrahydroisoquinoline-2-carboxylate hydrochloride

  • MDL number MFCD31811032
  • Cas Number : 2305252-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6750886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6750886
Availability details for
Quantity
  • Availability
  • Add to Cart