ENAH38A2CC60 Enamine

{6,6-difluorospiro[3.3]heptan-2-yl}methanesulfonyl chloride

  • MDL number MFCD31811034
  • Cas Number : 2305251-94-7
  • Molecular Weight : 244.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6750943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6750943
Availability details for
Quantity
  • Availability
  • Add to Cart