ENAH38A2CC67 Enamine

tert-butyl 3-(3-amino-2-oxopyrrolidin-1-yl)azetidine-1-carboxylate

  • MDL number MFCD31810807
  • Cas Number : 2305253-99-8
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6754277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754277
Availability details for
Quantity
  • Availability
  • Add to Cart