ENAH38A2CC6A Enamine

4-(dimethylphosphoryl)benzene-1,2-diamine

  • MDL number MFCD31810813
  • Cas Number : 2305255-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6754541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754541
Availability details for
Quantity
  • Availability
  • Add to Cart