ENAH38A2CC6D Enamine

tert-butyl 3-(chloromethyl)-3-methoxyazetidine-1-carboxylate

  • MDL number MFCD31810819
  • Cas Number : 2305252-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758591

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6758591
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart