ENAH38A2CC78 Enamine

tert-butyl 2-fluoromorpholine-4-carboxylate

  • MDL number MFCD31810876
  • Cas Number : 2305254-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758790
Availability details for
Quantity
  • Availability
  • Add to Cart